Assay Details
Target Gene Details
Entrez Gene ID: | 708 |
Gene Name: | complement C1q binding protein |
Gene Aliases: |
GC1QBP, HABP1, SF2p32, gC1Q-R, gC1qR, p32 |
Location: |
Chr.17:5432779-5439151 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QBP | NM_001212.3 | NP_001203.1 | ||
| AF275902.1 | AAF78763.1 | |||
| AK291516.1 | ||||
| BC000435.2 | AAH00435.1 | |||
| BC013731.1 | AAH13731.1 | |||
| L04636.1 | AAA16315.1 |
Target Gene Details
Entrez Gene ID: | 84268 |
Gene Name: | RPA interacting protein |
Gene Aliases: |
HRIP, RIP |
Location: |
Chr.17:5419185-5433020 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPAIN | NM_001033002.3 | 7 | 1405 | NP_001028174.2 |
| NM_001160244.1 | 6 | 1264 | NP_001153716.1 | |
| NM_001160246.1 | 4 | 1088 | NP_001153718.1 | |
| NM_001160266.1 | 5 | 1200 | NP_001153738.1 | |
| NR_027682.1 | 6 | 1341 | ||
| NR_027683.1 | 6 | 1293 | ||
| NR_027684.1 | 5 | 1152 | ||
| NR_027685.1 | 5 | 1229 | ||
| AA745783.1 | 1 | 136 | ||
| AK098491.1 | 2 | 1807 | BAC05317.1 | |
| AK226044.1 | 4 | 844 | ||
| AY775314.1 | 7 | 1405 | AAX14368.1 | |
| AY775315.1 | 6 | 1341 | AAX14369.1 | |
| AY775316.1 | 6 | 1264 | AAX14370.1 | |
| AY775317.1 | 6 | 1558 | AAX14371.1 | |
| AY775318.1 | 5 | 1417 | AAX14372.1 | |
| AY775319.1 | 5 | 1152 | AAX14373.1 | |
| AY775320.1 | 5 | 1229 | AAX14374.1 | |
| AY775321.1 | 4 | 1088 | AAX14375.1 | |
| AY775323.1 | 6 | 1293 | AAX14377.1 | |
| BC046349.1 | AAH46349.1 | |||
| BC051849.2 | AAH51849.1 | |||
| BG059980.1 | 1 | 130 | ||
| BI091755.1 | 4 | 524 | ||
| CK005117.1 | 4 | 547 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map