Assay Details
Target Gene Details
Entrez Gene ID: | 79590 |
Gene Name: | mitochondrial ribosomal protein L24 |
Gene Aliases: |
L24mt, MRP-L18, MRP-L24 |
Location: |
Chr.1:156737302-156741267 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL24 | NM_024540.3 | NP_078816.2 | ||
| NM_145729.2 | NP_663781.1 | |||
| XM_011509981.2 | XP_011508283.1 | |||
| XM_011509982.2 | XP_011508284.1 | |||
| AK024570.1 | BAB14929.1 | |||
| AK222500.1 | BAD96220.1 | |||
| AK223066.1 | BAD96786.1 | |||
| BC012440.1 | AAH12440.1 | |||
| BC016700.1 | AAH16700.1 | |||
| BG706700.1 | ||||
| CR457329.1 | CAG33610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| esv3578200 | Chr.1:156731053 - 156740837 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map