Assay Details
Target Gene Details
Entrez Gene ID: | 8697 |
Gene Name: | cell division cycle 23 |
Gene Aliases: |
ANAPC8, APC8, CUT23 |
Location: |
Chr.5:138187648-138213343 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC23 | NM_004661.3 | NP_004652.2 | ||
| AB011472.1 | BAA75628.1 | |||
| AF053977.1 | AAC70920.1 | |||
| AF191341.1 | AAF05755.1 | |||
| AK223250.1 | BAD96970.1 | |||
| AK291610.1 | ||||
| AK304635.1 | ||||
| BC017713.1 | AAH17713.1 | |||
| BT009810.1 | AAP88812.1 |
More Information
Set Membership: |
|

Genomic Map