Assay Details
Target Gene Details
Entrez Gene ID: | 23348 |
Gene Name: | dedicator of cytokinesis 9 |
Gene Aliases: |
ZIZ1, ZIZIMIN1 |
Location: |
Chr.13:98793487-99086406 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 54 - Exon 54 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038016 | Chr.13:98665177 - 98817733 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map