Assay Details
Target Gene Details
Entrez Gene ID: | 4171 |
Gene Name: | minichromosome maintenance complex component 2 |
Gene Aliases: |
BM28, CCNL1, CDCL1, D3S3194, DFNA70, MITOTIN, cdc19 |
Location: |
Chr.3:127598357-127622436 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM2 | NM_004526.3 | NP_004517.2 | ||
| NR_073375.1 | ||||
| AK128291.1 | ||||
| AK299934.1 | ||||
| AK304204.1 | ||||
| AK309335.1 | ||||
| BC006165.1 | ||||
| BC007670.2 | AAH07670.2 | |||
| BC007938.2 | AAH07938.2 | |||
| BC014272.2 | AAH14272.2 | |||
| BC017258.2 | AAH17258.2 | |||
| BC017490.1 | AAH17490.2 | |||
| BC030131.2 | AAH30131.2 | |||
| BT009734.1 | AAP88736.1 | |||
| D21063.1 | ||||
| D83987.1 | BAA12177.1 | |||
| X67334.1 | CAA47749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8563n54 | Chr.3:127543914 - 127685471 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map