Assay Details
Target Gene Details
Entrez Gene ID: | 26056 |
Gene Name: | RAB11 family interacting protein 5 |
Gene Aliases: |
GAF1, RIP11, pp75 |
Location: |
Chr.2:73073382-73113018 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11FIP5 | NM_015470.2 | 5 | 3891 | NP_056285.1 |
| XM_005264251.3 | 6 | 5527 | XP_005264308.1 | |
| XM_005264252.3 | 6 | 5845 | XP_005264309.1 | |
| XM_006711985.3 | 7 | 5620 | XP_006712048.1 | |
| XM_011532753.2 | 7 | 5130 | XP_011531055.1 | |
| XM_017003788.1 | 7 | 5182 | XP_016859277.1 | |
| AB020664.1 | 5 | 3891 | ||
| AF334812.1 | 5 | 3890 | AAK20892.1 | |
| AK023823.1 | 4 | 3556 | ||
| AK024727.1 | 1 | 1343 | ||
| AL117579.1 | 3 | 2451 | CAB56002.1 | |
| BC035013.1 | 5 | 3814 | AAH35013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002295 | Chr.2:72989026 - 73123263 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116914899] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map