Assay Details
Target Gene Details
Entrez Gene ID: | 125 |
Gene Name: | alcohol dehydrogenase 1B (class I), beta polypeptide |
Gene Aliases: |
ADH2, HEL-S-117 |
Location: |
Chr.4:99306387-99321442 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH1B | NM_000668.5 | 9 | 1628 | NP_000659.2 |
| NM_001286650.1 | 10 | 1750 | NP_001273579.1 | |
| AF153821.1 | 9 | 1585 | AAD37446.1 | |
| AK316501.1 | ||||
| BC033009.2 | 9 | 1587 | AAH33009.1 | |
| BX647469.1 | 9 | 3612 | ||
| D00137.1 | 9 | 1537 | BAA00084.1 | |
| FJ224285.1 | 9 | 1587 | ||
| M21692.1 | 9 | 1549 | AAA51592.1 | |
| M24317.1 | 9 | 1547 | AAA51884.1 | |
| X03350.1 | 9 | 1589 | CAA27056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain |
|
| esv2659036 | Chr.4:99284003 - 99344468 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map