Assay Details
Target Gene Details
Entrez Gene ID: | 115350 |
Gene Name: | Fc receptor like 1 |
Gene Aliases: |
CD307a, FCRH1, IFGP1, IRTA5 |
Location: |
Chr.1:157792249-157820150 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587657 | Chr.1:157793395 - 157801763 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map