Assay Details
Target Gene Details
Entrez Gene ID: | 84941 |
Gene Name: | hematopoietic SH2 domain containing |
Gene Aliases: |
ALX, HSH2 |
Location: |
Chr.19:16134028-16158575 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSH2D | NM_001291274.1 | 6 | 1315 | NP_001278203.1 |
| NM_032855.3 | NP_116244.1 | |||
| NR_111903.1 | 6 | 793 | ||
| NR_111904.1 | 5 | 1183 | ||
| XM_011528386.2 | XP_011526688.1 | |||
| XM_011528387.2 | 4 | 379 | XP_011526689.1 | |
| XM_011528388.2 | XP_011526690.1 | |||
| XM_011528389.2 | 4 | 403 | XP_011526691.1 | |
| XM_011528390.1 | XP_011526692.1 | |||
| XM_017027402.1 | XP_016882891.1 | |||
| XM_017027403.1 | XP_016882892.1 | |||
| XM_017027404.1 | XP_016882893.1 | |||
| AK027792.1 | BAB55372.1 | |||
| AK131222.1 | 4 | 895 | BAD18408.1 | |
| AK298682.1 | 3 | 726 | ||
| AK300027.1 | 3 | 739 | ||
| AK307563.1 | 4 | 336 | ||
| AY319652.1 | AAQ81285.1 | |||
| BC016826.1 | AAH16826.1 | |||
| BC025237.2 | AAH25237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643791 | Chr.19:16149438 - 16170044 on Build GRCh38 | Loss |
|
| nsv833767 | Chr.19:16121391 - 16347760 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map