Assay Details
Target Gene Details
Entrez Gene ID: | 81555 |
Gene Name: | Yip1 domain family member 5 |
Gene Aliases: |
FinGER5, SB140, SMAP-5, SMAP5, YIP1A |
Location: |
Chr.5:144158159-144170714 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF5 | NM_001024947.3 | 6 | 3080 | NP_001020118.1 |
| NM_001271732.1 | 5 | 2960 | NP_001258661.1 | |
| NM_030799.8 | 6 | 2829 | NP_110426.4 | |
| AA811265.1 | 1 | 384 | ||
| AB014733.1 | 2 | 2100 | BAB20270.1 | |
| AY037152.1 | AAK67644.1 | |||
| AY640926.1 | 6 | 2745 | AAV51257.1 | |
| AY640934.1 | AAV51260.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115282675] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map