Assay Details
Target Gene Details
Entrez Gene ID: | 11012 |
Gene Name: | kallikrein related peptidase 11 |
Gene Aliases: |
PRSS20, TLSP |
Location: |
Chr.19:51022231-51028034 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK11 | NM_001167605.1 | NP_001161077.1 | ||
| NM_006853.2 | NP_006844.1 | |||
| NM_144947.1 | NP_659196.1 | |||
| XM_005258439.3 | XP_005258496.1 | |||
| XM_011526369.1 | XP_011524671.1 | |||
| AB012917.1 | BAA33404.1 | |||
| AB013730.1 | BAA88713.1 | |||
| AB041036.1 | BAA96797.1 | |||
| AY359014.1 | AAQ89373.1 | |||
| BC022068.1 | AAH22068.1 |
More Information
Set Membership: |
|

Genomic Map