Assay Details
Target Gene Details
Entrez Gene ID: | 1020 |
Gene Name: | cyclin dependent kinase 5 |
Gene Aliases: |
LIS7, PSSALRE |
Location: |
Chr.7:151053812-151057965 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK5 | NM_001164410.2 | NP_001157882.1 | ||
| NM_004935.3 | NP_004926.1 | |||
| AK026533.1 | ||||
| AY049778.1 | AAL15435.1 | |||
| AY927560.1 | ||||
| BC005115.1 | AAH05115.1 | |||
| BT006680.1 | AAP35326.1 | |||
| BU527089.1 | ||||
| CA429180.1 | ||||
| CR457041.1 | CAG33322.1 | |||
| DQ411039.1 | ||||
| KU177968.1 | ||||
| KU177969.1 | ||||
| L04658.1 | ||||
| X66364.1 | CAA47007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831188 | Chr.7:151038117 - 151215789 on Build GRCh38 | Loss |
|
| nsv526410 | Chr.7:151017725 - 151078219 on Build GRCh38 | Loss |
|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
| nsv524928 | Chr.7:151034611 - 151069712 on Build GRCh38 | Gain |
|
| dgv6756n100 | Chr.7:151033912 - 151093430 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map