Assay Details
Target Gene Details
Entrez Gene ID: | 3921 |
Gene Name: | ribosomal protein SA |
Gene Aliases: |
37LRP, 67LR, ICAS, LAMBR, LAMR1, LBP, LBP/p40, LRP, LRP/LR, NEM/1CHD4, SA, lamR, p40 |
Location: |
Chr.3:39406689-39412542 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPSA | NM_001304288.1 | NP_001291217.1 | ||
| NM_002295.5 | NP_002286.2 | |||
| BC066941.1 | AAH66941.1 | |||
| BC070263.1 | AAH70263.1 | |||
| BT007219.1 | AAP35883.1 | |||
| J03799.1 | AAA36161.1 | |||
| S37431.1 | AAB22299.1 | |||
| X15005.1 | CAA33112.1 | |||
| X61156.1 | CAA43469.1 |
Target Gene Details
Entrez Gene ID: | 574040 |
Gene Name: | small nucleolar RNA, H/ACA box 6 |
Gene Aliases: |
ACA6 |
Location: |
Chr.3:39408391-39408539 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNORA6 | NR_002325.1 | |||
| AF375472.1 | ||||
| AJ609429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv963310 | Chr.3:39408597 - 39412433 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map