Assay Details
Target Gene Details
Entrez Gene ID: | 54986 |
Gene Name: | unc-51 like kinase 4 |
Gene Aliases: |
FAM7C1, REC01035 |
Location: |
Chr.3:41246599-41963020 on Build GRCh38 |
Assay Gene Location: | Within Intron 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ULK4 | NM_001322501.1 | NP_001309430.1 | ||
| NM_017886.3 | NP_060356.2 | |||
| NR_136342.1 | ||||
| XM_011533880.2 | XP_011532182.1 | |||
| XM_017006711.1 | XP_016862200.1 | |||
| AK093396.1 | ||||
| AK304341.1 | ||||
| AL133104.1 | CAB61411.1 | |||
| BC040739.1 | AAH40739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521572 | Chr.3:41286691 - 41318043 on Build GRCh38 | Loss |
|
| nsv1002077 | Chr.3:41269871 - 41303354 on Build GRCh38 | Loss |
|
| nsv3779 | Chr.3:41292209 - 41809712 on Build GRCh38 | Deletion |
|
| esv2759143 | Chr.3:41297661 - 41732261 on Build GRCh38 | Loss |
|
| nsv834670 | Chr.3:41297663 - 41444927 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map