Assay Details
Target Gene Details
Entrez Gene ID: | 23231 |
Gene Name: | SEL1L family member 3 |
Gene Aliases: |
Sel-1L3 |
Location: |
Chr.4:25697790-25863595 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEL1L3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893899 | Chr.4:25652944 - 25747681 on Build GRCh38 | Loss |
|
| esv2759236 | Chr.4:25704199 - 25738584 on Build GRCh38 | Loss |
|
| esv2762416 | Chr.4:25712604 - 25713093 on Build GRCh38 | Loss |
|
| esv2676250 | Chr.4:25710333 - 25734252 on Build GRCh38 | Deletion |
|
| nsv997544 | Chr.4:25362237 - 25915432 on Build GRCh38 | Gain |
|
| esv3599973 | Chr.4:25710346 - 25734271 on Build GRCh38 | Loss |
|

Genomic Map