Assay Details
Target Gene Details
Entrez Gene ID: | 23231 |
Gene Name: | SEL1L family member 3 |
Gene Aliases: |
Sel-1L3 |
Location: |
Chr.4:25697790-25863595 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEL1L3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv980188 | Chr.4:25713016 - 25721336 on Build GRCh38 | Duplication |
|
| esv3893899 | Chr.4:25652944 - 25747681 on Build GRCh38 | Loss |
|
| esv2759236 | Chr.4:25704199 - 25738584 on Build GRCh38 | Loss |
|
| nsv441864 | Chr.4:25712604 - 25729254 on Build GRCh38 | Loss |
|
| nsv822507 | Chr.4:25713318 - 25731460 on Build GRCh38 | Loss |
|
| esv2676250 | Chr.4:25710333 - 25734252 on Build GRCh38 | Deletion |
|
| nsv997544 | Chr.4:25362237 - 25915432 on Build GRCh38 | Gain |
|
| esv3599973 | Chr.4:25710346 - 25734271 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74943665] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map