Assay Details
Target Gene Details
Entrez Gene ID: | 644150 |
Gene Name: | WAS/WASL interacting protein family member 3 |
Gene Aliases: |
CR16 |
Location: |
Chr.7:29806554-29917066 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WIPF3 | NM_001080529.2 | NP_001073998.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830936 | Chr.7:29733892 - 29960517 on Build GRCh38 | Gain |
|
| esv2759521 | Chr.7:29608737 - 29895756 on Build GRCh38 | Gain |
|
| nsv1019527 | Chr.7:29722658 - 30413302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map