Assay Details
Target Gene Details
Entrez Gene ID: | 100507428 |
Gene Name: | long intergenic non-protein coding RNA 458 |
Gene Aliases: |
LncRNA-ES3 |
Location: |
Chr.13:54124324-54132871 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00458 | NR_108062.1 | |||
| NR_108063.1 | 3 | 844 | ||
| CX163049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759938 | Chr.13:54117897 - 54137101 on Build GRCh38 | Loss |
|
| esv3892347 | Chr.13:53543177 - 54476363 on Build GRCh38 | Gain |
|
| nsv1049 | Chr.13:54110219 - 54154846 on Build GRCh38 | Deletion |
|

Genomic Map