Assay Details
Target Gene Details
| Entrez Gene ID: | 54715 | 
| Gene Name: | RNA binding protein, fox-1 homolog 1 | 
| Gene Aliases: | 2BP1, A2BP1, FOX-1, FOX1, HRNBP1 | 
| Location: | Chr.16:5239749-7713343 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| RBFOX1 | XM_017023319.1 | XP_016878808.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1723 | Chr.16:5322712 - 5339004 on Build GRCh38 | Insertion |  RBFOX1 | 
| nsv1035156 | Chr.16:5091032 - 5996934 on Build GRCh38 | Gain |  MIR8065  RBFOX1  LINC01570  EEF2KMT | 
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |  RBFOX1  DNAJA3  SEC14L5  MIR6769A  MGRN1  CORO7  CORO7-PAM16  SEPT12  ZNF500  TFAP4  GLIS2-AS1  EEF2KMT  ADCY9  SMIM22  GLIS2  VASN  C16orf96  UBALD1  ROGDI  LOC105371063  LINC01569  ANKS3  UBN1  NAGPA-AS1  HMOX2  PAM16  ALG1  C16orf89  NAGPA  NUDT16L1  SRL  C16orf71  PPL  NMRAL1  GLYR1  CDIP1 | 
| nsv1045304 | Chr.16:5205761 - 5326875 on Build GRCh38 | Loss |  RBFOX1 | 
| dgv2698n100 | Chr.16:5304641 - 5327717 on Build GRCh38 | Loss |  RBFOX1 | 
| nsv1036241 | Chr.16:5271882 - 5387609 on Build GRCh38 | Loss |  RBFOX1 | 
| esv3892764 | Chr.16:5316436 - 5404485 on Build GRCh38 | Loss |  RBFOX1 | 
| esv2760069 | Chr.16:4922849 - 5374828 on Build GRCh38 | Gain+Loss |  RBFOX1  SEC14L5  PPL  NAGPA-AS1  ALG1  EEF2KMT  C16orf89  NAGPA | 
| nsv428318 | Chr.16:5011594 - 5374828 on Build GRCh38 | Gain |  RBFOX1  SEC14L5  NAGPA-AS1  ALG1  EEF2KMT  C16orf89  NAGPA | 
| esv2762021 | Chr.16:5293419 - 5377238 on Build GRCh38 | Loss |  RBFOX1 | 
| nsv1037303 | Chr.16:5287645 - 5327595 on Build GRCh38 | Loss |  RBFOX1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Molecular Function -
 RNA metabolism protein
								
								   RNA metabolism protein
								
							
								 RNA processing factor
								
								   RNA processing factor
								
							
								 RNA splicing factor
								
								   RNA splicing factor
								
							
								 nucleic acid metabolism protein
								
								   nucleic acid metabolism protein
								
							
						
					Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top