Assay Details
Target Gene Details
Entrez Gene ID: | 5590 |
Gene Name: | protein kinase C zeta |
Gene Aliases: |
PKC-ZETA, PKC2 |
Location: |
Chr.1:2050470-2185395 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCZ | NM_002744.4 | NP_002735.3 | ||
| XM_011541773.1 | XP_011540075.1 | |||
| XM_011541775.2 | XP_011540077.1 | |||
| XM_017001789.1 | XP_016857278.1 | |||
| XM_017001790.1 | XP_016857279.1 | |||
| XM_017001803.1 | XP_016857292.1 | |||
| AK290995.1 | ||||
| BC008058.2 | AAH08058.1 | |||
| BC014270.2 | AAH14270.1 | |||
| BT007082.1 | AAP35745.1 | |||
| L14283.1 | AAA36488.1 | |||
| Z15108.1 | CAA78813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951425 | Chr.1:2018162 - 2112261 on Build GRCh38 | Deletion |
|
| nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
|
| nsv834446 | Chr.1:1960604 - 2156435 on Build GRCh38 | Loss |
|
| esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map