Assay Details
Target Gene Details
Entrez Gene ID: | 23266 |
Gene Name: | adhesion G protein-coupled receptor L2 |
Gene Aliases: |
CIRL2, CL2, LEC1, LPHH1, LPHN2 |
Location: |
Chr.1:81306155-81992738 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL2 | XM_017000782.1 | XP_016856271.1 | ||
| AK123422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750840 | Chr.1:79603397 - 81378201 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map