Assay Details
Target Gene Details
Entrez Gene ID: | 353500 |
Gene Name: | bone morphogenetic protein 8a |
Gene Aliases: |
- |
Location: |
Chr.1:39491626-39529869 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMP8A | NM_181809.3 | NP_861525.2 | ||
| XM_006710616.3 | XP_006710679.1 | |||
| XM_011541381.2 | XP_011539683.1 | |||
| XM_011541382.2 | XP_011539684.1 | |||
| XM_017001198.1 | XP_016856687.1 | |||
| AY303954.1 | AAP74559.1 | |||
| BC131617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829649 | Chr.1:39505283 - 39638842 on Build GRCh38 | Loss |
|
| nsv508160 | Chr.1:39503058 - 39564874 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map