Assay Details
Target Gene Details
Entrez Gene ID: | 200150 |
Gene Name: | phospholipase D family member 5 |
Gene Aliases: |
PLDC |
Location: |
Chr.1:242085051-242530546 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549462 | Chr.1:242037945 - 242392647 on Build GRCh38 | Gain |
|
| dgv16e55 | Chr.1:241861796 - 242169657 on Build GRCh38 | Gain |
|
| nsv528016 | Chr.1:242060097 - 242288048 on Build GRCh38 | Gain |
|
| nsv1002309 | Chr.1:241878940 - 242132333 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map