Assay Details
Target Gene Details
Entrez Gene ID: | 9563 |
Gene Name: | hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase |
Gene Aliases: |
CORTRD1, G6PDH, GDH |
Location: |
Chr.1:9234767-9271337 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H6PD | NM_001282587.1 | NP_001269516.1 | ||
NM_004285.3 | NP_004276.2 | |||
XM_005263540.4 | XP_005263597.1 | |||
XM_006711052.3 | XP_006711115.1 | |||
XM_017002865.1 | XP_016858354.1 | |||
XM_017002866.1 | XP_016858355.1 | |||
AJ012590.1 | CAA10071.1 | |||
BC081559.1 | AAH81559.1 | |||
CR749282.1 | CAH18137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv40e59 | Chr.1:9257906 - 9261004 on Build GRCh38 | Duplication |
![]() |
dgv2e55 | Chr.1:9255788 - 9344634 on Build GRCh38 | Gain |
![]() ![]() |
nsv545350 | Chr.1:9258342 - 9306793 on Build GRCh38 | Gain |
![]() ![]() |
dgv16n100 | Chr.1:9237601 - 9351410 on Build GRCh38 | Gain |
![]() ![]() |
nsv1128586 | Chr.1:9258241 - 9260741 on Build GRCh38 | Duplication |
![]() |
nsv1075364 | Chr.1:9258040 - 9260741 on Build GRCh38 | Duplication |
![]() |
nsv832092 | Chr.1:9126509 - 9304543 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv97n54 | Chr.1:9249687 - 9352728 on Build GRCh38 | Gain |
![]() ![]() |
dgv12e212 | Chr.1:9243684 - 9344908 on Build GRCh38 | Gain |
![]() ![]() |
esv2763409 | Chr.1:9247978 - 9344958 on Build GRCh38 | Gain |
![]() ![]() |
esv23318 | Chr.1:9257747 - 9261167 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
