Assay Details
Target Gene Details
| Entrez Gene ID: | 148398 | 
| Gene Name: | sterile alpha motif domain containing 11 | 
| Gene Aliases: | MRS | 
| Location: | Chr.1:925741-944581 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 4 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| SAMD11 | NM_152486.2 | NP_689699.2 | ||
| CN293867.1 | ||||
| CN342795.1 | ||||
| JX093066.1 | ||||
| JX093067.1 | ||||
| JX093068.1 | ||||
| JX093069.1 | ||||
| JX093070.1 | ||||
| JX093071.1 | ||||
| JX093072.1 | ||||
| JX093073.1 | ||||
| JX093074.1 | ||||
| JX093075.1 | ||||
| JX093076.1 | ||||
| JX093077.1 | ||||
| JX093078.1 | ||||
| JX093079.1 | ||||
| JX093080.1 | ||||
| JX093081.1 | ||||
| JX093082.1 | ||||
| JX093083.1 | ||||
| JX093084.1 | ||||
| JX093085.1 | ||||
| JX093086.1 | ||||
| JX093087.1 | ||||
| JX093088.1 | ||||
| JX093089.1 | ||||
| JX093090.1 | ||||
| JX093091.1 | ||||
| JX093092.1 | ||||
| JX093093.1 | ||||
| JX093094.1 | ||||
| JX093095.1 | ||||
| JX093096.1 | ||||
| JX093097.1 | ||||
| JX093098.1 | ||||
| JX093099.1 | ||||
| JX093100.1 | ||||
| JX093101.1 | ||||
| JX093102.1 | ||||
| JX093103.1 | ||||
| JX093104.1 | ||||
| JX093105.1 | ||||
| JX093106.1 | ||||
| JX093107.1 | ||||
| JX093108.1 | ||||
| JX093109.1 | ||||
| JX093110.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2762302 | Chr.1:914637 - 1106320 on Build GRCh38 | Gain |  LOC284600  AGRN  RNF223  PLEKHN1  PERM1  SAMD11  HES4  LOC105378948  ISG15  KLHL17  LOC100288175  LOC100130417  NOC2L  LOC107985728  C1orf159 | 
| nsv1142736 | Chr.1:933720 - 934720 on Build GRCh38 | Deletion |  SAMD11 | 
| nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |  TMEM88B  DVL1  VWA1  TNFRSF4  SLC35E2B  MIR6859-2  ATAD3A  NOC2L  WASH7P  TMEM52  FNDC10  LOC105378592  LINC00115  MXRA8  MIR1302-2  FAM87B  LOC105378947  KLHL17  MIR429  TNFRSF18  LOC100288069  CPSF3L  FAM41C  ATAD3B  LOC105378948  B3GALT6  MIR6727  MMP23A  ACAP3  CCNL2  LOC100129534  LOC107984841  LOC100132287  SCNN1D  PRKCZ  RNF223  LOC101928626  PLEKHN1  GNB1  LOC105378589  NADK  FAAP20  TMEM240  FAM132A  MORN1  CDK11A  SSU72  LOC100134822  LOC148413  TTLL10-AS1  LOC105378591  LOC729737  ISG15  AURKAIP1  LOC100288175  DDX11L1  LOC102724312  UBE2J2  MRPL20  GABRD  LINC01128  MIB2  CALML6  OR4F5  PERM1  SLC35E2  C1orf159  CFAP74  MMP23B  LOC100133331  TAS1R3  MIR200A  MIR6808  SDF4  ATAD3C  PUSL1  HES4  MIR200B  LOC107985729  MTND1P23  LOC100130417  MIR6723  LOC107985728  LOC105378949  LOC284600  AGRN  TTLL10  SKI  CPTP  LOC100287934  MIR6859-1  MIR6726  LOC100506504  SAMD11  OR4F29  MTND2P28  CDK11B  LOC102725121  FAM138A  OR4F16  ANKRD65  LINC01342 | 
| nsv544904 | Chr.1:929558 - 934877 on Build GRCh38 | Gain |  SAMD11 | 
| nsv950451 | Chr.1:899421 - 1094520 on Build GRCh38 | Deletion |  LOC284600  AGRN  RNF223  PLEKHN1  PERM1  SAMD11  HES4  LOC105378948  ISG15  KLHL17  LOC100288175  LOC100130417  NOC2L  LOC107985728  C1orf159 | 
| nsv509035 | Chr.1:891406 - 985724 on Build GRCh38 | Insertion |  PERM1  LOC284600  SAMD11  KLHL17  LOC100130417  NOC2L  LOC107985728  PLEKHN1 | 
| dgv9n54 | Chr.1:925754 - 952840 on Build GRCh38 | Loss |  SAMD11  NOC2L | 
| nsv428334 | Chr.1:874371 - 1220569 on Build GRCh38 | Gain |  MIR429  TNFRSF18  MIR200A  TNFRSF4  SDF4  TTLL10-AS1  FAM41C  HES4  LOC105378948  ISG15  MIR200B  LOC100288175  LOC100130417  NOC2L  LOC107985728  LOC284600  AGRN  TTLL10  RNF223  PLEKHN1  PERM1  SAMD11  KLHL17  C1orf159  LINC01342 | 
| nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss |  TMEM88B  TMEM240  DVL1  VWA1  TNFRSF4  FAM132A  CDK11A  SSU72  LOC148413  TTLL10-AS1  SLC35E2B  ISG15  AURKAIP1  LOC100288175  LOC102724312  ATAD3A  NOC2L  UBE2J2  MRPL20  FNDC10  LINC01128  MIB2  LINC00115  MXRA8  PERM1  SLC35E2  FAM87B  KLHL17  C1orf159  MMP23B  MIR429  TAS1R3  TNFRSF18  MIR200A  MIR6808  SDF4  LOC100288069  CPSF3L  FAM41C  ATAD3C  ATAD3B  PUSL1  HES4  LOC105378948  B3GALT6  MIR6727  MMP23A  MIR200B  LOC107985729  LOC100130417  LOC107985728  ACAP3  CCNL2  LOC284600  AGRN  TTLL10  SCNN1D  CPTP  RNF223  LOC100287934  MIR6726  PLEKHN1  SAMD11  NADK  CDK11B  ANKRD65  LINC01342 | 
| dgv5n100 | Chr.1:585989 - 1114424 on Build GRCh38 | Gain |  LOC100288069  FAM41C  HES4  LOC105378948  ISG15  MTND1P23  LOC100288175  LOC100130417  MIR6723  NOC2L  LOC107985728  LOC284600  LOC107984841  LINC01128  AGRN  LINC00115  RNF223  LOC100287934  LOC101928626  PLEKHN1  PERM1  SAMD11  FAM87B  LOC105378947  MTND2P28  KLHL17  OR4F16  C1orf159  LOC100133331 | 
| nsv517709 | Chr.1:817186 - 1275912 on Build GRCh38 | Gain+Loss |  MIR429  TNFRSF18  MIR200A  TNFRSF4  FAM132A  SDF4  TTLL10-AS1  FAM41C  HES4  LOC105378948  B3GALT6  ISG15  MIR200B  LOC100288175  LOC100130417  NOC2L  LOC107985728  UBE2J2  LOC284600  LINC01128  AGRN  TTLL10  LINC00115  RNF223  PLEKHN1  PERM1  SAMD11  FAM87B  KLHL17  C1orf159  LINC01342 | 
| dgv1n111 | Chr.1:690090 - 939522 on Build GRCh38 | Duplication |  LOC284600  LOC107984841  LINC01128  LINC00115  LOC100287934  LOC100288069  FAM41C  SAMD11  FAM87B  OR4F16  LOC100130417  LOC107985728  LOC100133331 | 
| nsv1074938 | Chr.1:933719 - 935120 on Build GRCh38 | Deletion |  SAMD11 | 
| dgv3n111 | Chr.1:783283 - 939522 on Build GRCh38 | Duplication |  LOC284600  SAMD11  FAM87B  LINC01128  LINC00115  LOC100130417  LOC100287934  LOC107985728  FAM41C | 
| dgv6n54 | Chr.1:917392 - 934930 on Build GRCh38 | Loss |  LOC284600  SAMD11  LOC100130417  LOC107985728 | 
| dgv2n67 | Chr.1:877618 - 1426500 on Build GRCh38 | Gain |  MIR429  TAS1R3  TMEM88B  TNFRSF18  DVL1  MIR200A  MIR6808  TNFRSF4  FAM132A  SDF4  LOC148413  CPSF3L  TTLL10-AS1  PUSL1  HES4  LOC105378948  B3GALT6  MIR6727  ISG15  AURKAIP1  MIR200B  LOC100288175  LOC100130417  NOC2L  LOC107985728  ACAP3  CCNL2  UBE2J2  MRPL20  LOC284600  AGRN  TTLL10  SCNN1D  CPTP  RNF223  MXRA8  MIR6726  PLEKHN1  PERM1  SAMD11  KLHL17  ANKRD65  C1orf159  LINC01342 | 
| nsv544898 | Chr.1:924665 - 943937 on Build GRCh38 | Gain |  SAMD11  LOC107985728 | 
| nsv832980 | Chr.1:848279 - 1007037 on Build GRCh38 | Gain |  PERM1  LOC284600  SAMD11  HES4  LINC01128  KLHL17  LOC100130417  NOC2L  LOC107985728  PLEKHN1  FAM41C | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	For this assay, SNP(s) [rs115552829,rs116296342] are located under a primer or probe  sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top