Assay Details
Target Gene Details
Entrez Gene ID: | 55268 |
Gene Name: | enoyl-CoA hydratase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.1:52895910-52927212 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECHDC2 | XM_011541709.2 | XP_011540011.1 | ||
| XM_011541710.2 | XP_011540012.1 | |||
| XM_011541711.2 | XP_011540013.1 | |||
| XM_011541712.2 | XP_011540014.1 | |||
| XM_011541721.2 | XP_011540023.1 | |||
| XM_011541724.2 | XP_011540026.1 | |||
| XM_011541725.2 | XP_011540027.1 | |||
| XM_017001638.1 | XP_016857127.1 | |||
| XM_017001641.1 | XP_016857130.1 | |||
| AK302285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
| nsv829859 | Chr.1:52723470 - 52925273 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map