Assay Details
Target Gene Details
Entrez Gene ID: | 11146 |
Gene Name: | glomulin, FKBP associated protein |
Gene Aliases: |
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM |
Location: |
Chr.1:92245786-92441936 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLMN | XM_005270401.3 | 18 | 2243 | XP_005270458.1 |
| XM_006710309.2 | 19 | 2383 | XP_006710372.1 | |
| XM_011540546.2 | 21 | 2544 | XP_011538848.1 | |
| XM_017000137.1 | 19 | 2749 | XP_016855626.1 | |
| XM_017000138.1 | 18 | 2707 | XP_016855627.1 | |
| XM_017000139.1 | 18 | 2644 | XP_016855628.1 | |
| XM_017000140.1 | 18 | 2359 | XP_016855629.1 | |
| XM_017000141.1 | 18 | 2276 | XP_016855630.1 | |
| XM_017000142.1 | 18 | 2341 | XP_016855631.1 | |
| XM_017000143.1 | 18 | 2722 | XP_016855632.1 | |
| XM_017000144.1 | 18 | 2267 | XP_016855633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830592 | Chr.1:92203725 - 92340072 on Build GRCh38 | Gain |
|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map