Assay Details
Target Gene Details
Entrez Gene ID: | 9651 |
Gene Name: | phospholipase C eta 2 |
Gene Aliases: |
PLC-L4, PLC-eta2, PLCL4, PLCeta2 |
Location: |
Chr.1:2412481-2505530 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCH2 | XM_011542455.2 | XP_011540757.1 | ||
| XM_011542456.2 | XP_011540758.1 | |||
| XM_017002870.1 | XP_016858359.1 | |||
| XM_017002872.1 | XP_016858361.1 | |||
| XM_017002873.1 | XP_016858362.1 | |||
| XM_017002874.1 | XP_016858363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951557 | Chr.1:2266062 - 2564661 on Build GRCh38 | Deletion |
|
| dgv50n54 | Chr.1:2401592 - 2518955 on Build GRCh38 | Loss |
|
| nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80045357] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map