Assay Details
Target Gene Details
Entrez Gene ID: | 57540 |
Gene Name: | dispatched RND transporter family member 3 |
Gene Aliases: |
PTCHD2 |
Location: |
Chr.1:11479155-11537583 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DISP3 | NM_020780.1 | NP_065831.1 | ||
| XM_011541830.2 | XP_011540132.1 | |||
| XM_011541831.2 | XP_011540133.1 | |||
| XM_011541832.2 | XP_011540134.1 | |||
| XM_011541833.2 | XP_011540135.1 | |||
| XM_011541835.2 | XP_011540137.1 | |||
| AB037758.1 | ||||
| AB593117.1 | ||||
| AK311332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832536 | Chr.1:11328720 - 11502284 on Build GRCh38 | Loss |
|
| nsv2632 | Chr.1:11477020 - 11507795 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113875216,rs76452301] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map