Assay Details
Target Gene Details
Entrez Gene ID: | 100423027 |
Gene Name: | microRNA 4266 |
Gene Aliases: |
- |
Location: |
Chr.2:109313571-109313625 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4266 | NR_036224.1 |
Target Gene Details
Entrez Gene ID: | 5903 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2 |
Location: |
Chr.2:108719446-109842301 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP2 | XM_017004623.1 | XP_016860112.1 |
Target Gene Details
Entrez Gene ID: | 344558 |
Gene Name: | SH3 domain containing ring finger 3 |
Gene Aliases: |
POSH2, SH3MD4 |
Location: |
Chr.2:109129439-109504636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3RF3 | NM_001099289.2 | NP_001092759.1 | ||
| XM_011511109.2 | XP_011509411.1 | |||
| DQ307286.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28484 | Chr.2:109305606 - 109316939 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map