Assay Details
Target Gene Details
Entrez Gene ID: | 389072 |
Gene Name: | pleckstrin homology domain containing M3 |
Gene Aliases: |
DAPR, PLEKHM1L |
Location: |
Chr.2:207821288-208025675 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHM3 | NM_001080475.2 | NP_001073944.1 | ||
| XM_005246545.3 | XP_005246602.1 | |||
| XM_011511160.2 | XP_011509462.1 | |||
| XM_017004072.1 | XP_016859561.1 | |||
| XM_017004073.1 | XP_016859562.1 | |||
| BC151137.1 |
More Information
Set Membership: |
|

Genomic Map