Assay Details
Target Gene Details
Entrez Gene ID: | 57504 |
Gene Name: | metastasis associated 1 family member 3 |
Gene Aliases: |
- |
Location: |
Chr.2:42494569-42795032 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTA3 | NM_001282755.1 | NP_001269684.1 | ||
| XM_005264459.2 | XP_005264516.1 | |||
| AK127245.1 | ||||
| DA239202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583763 | Chr.2:42479558 - 42513741 on Build GRCh38 | Loss |
|
| nsv524679 | Chr.2:42350030 - 42576225 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map