Assay Details
Target Gene Details
Entrez Gene ID: | 200424 |
Gene Name: | tet methylcytosine dioxygenase 3 |
Gene Aliases: |
hCG_40738 |
Location: |
Chr.2:73982036-74108176 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TET3 | NM_001287491.1 | NP_001274420.1 | ||
| XM_005264187.3 | XP_005264244.1 | |||
| XM_011532682.2 | XP_011530984.1 | |||
| XM_011532683.2 | XP_011530985.1 | |||
| XM_011532684.2 | XP_011530986.1 | |||
| XM_011532685.2 | XP_011530987.1 | |||
| XM_011532686.2 | XP_011530988.1 | |||
| XM_011532687.2 | XP_011530989.1 | |||
| XM_011532688.2 | XP_011530990.1 | |||
| XM_011532689.2 | XP_011530991.1 | |||
| XM_011532690.1 | XP_011530992.1 | |||
| XM_017003566.1 | XP_016859055.1 | |||
| XM_017003567.1 | XP_016859056.1 | |||
| HQ220209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818048 | Chr.2:73998307 - 74085953 on Build GRCh38 | Gain |
|
| esv3584956 | Chr.2:73969982 - 74090008 on Build GRCh38 | Gain |
|
| nsv1011177 | Chr.2:73940216 - 74012778 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map