Assay Details
Target Gene Details
Entrez Gene ID: | 30818 |
Gene Name: | potassium voltage-gated channel interacting protein 3 |
Gene Aliases: |
CSEN, DREAM, KCHIP3 |
Location: |
Chr.2:95297324-95386077 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNIP3 | NM_013434.4 | NP_038462.1 | ||
| AB209076.1 | 2 | 827 | BAD92313.1 | |
| AF120102.1 | AAD20350.1 | |||
| AF199599.1 | AAF33684.1 | |||
| AF367022.1 | AAK53711.1 | |||
| AJ131730.2 | CAB56835.1 | |||
| AK315437.1 | ||||
| BC012850.1 | AAH12850.1 | |||
| BI755669.1 | ||||
| BT020075.1 | AAV38878.1 | |||
| DQ148485.1 | AAZ77802.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528011 | Chr.2:95265470 - 95391263 on Build GRCh38 | Loss |
|
| nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
|
| esv2759076 | Chr.2:95304390 - 95486811 on Build GRCh38 | Loss |
|
| nsv998014 | Chr.2:95247530 - 95630882 on Build GRCh38 | Gain |
|
| nsv953478 | Chr.2:95266753 - 95326752 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map