Assay Details
Target Gene Details
Entrez Gene ID: | 5903 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2 |
Location: |
Chr.2:108719446-109842301 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP2 | XM_017004623.1 | XP_016860112.1 |
Target Gene Details
Entrez Gene ID: | 344558 |
Gene Name: | SH3 domain containing ring finger 3 |
Gene Aliases: |
POSH2, SH3MD4 |
Location: |
Chr.2:109129439-109504636 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3RF3 | NM_001099289.2 | 1 | 235 | NP_001092759.1 |
| XM_011511109.2 | 1 | 337 | XP_011509411.1 | |
| DQ307286.1 | 1 | 100 |
Target Gene Details
Entrez Gene ID: | 100287216 |
Gene Name: | SH3RF3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:109127327-109130119 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3RF3-AS1 | NR_029193.1 | 1 | 345 | |
| AK297376.1 | 1 | 345 |
More Information
Set Membership: |
|

Genomic Map