Assay Details
Target Gene Details
Entrez Gene ID: | 114818 |
Gene Name: | kelch like family member 29 |
Gene Aliases: |
KBTBD9 |
Location: |
Chr.2:23385419-23708613 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL29 | NM_052920.1 | NP_443152.1 | ||
| XM_017003264.1 | XP_016858753.1 | |||
| BC013982.1 | AAH13982.1 | |||
| DR002067.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
|
| nsv581173 | Chr.2:23376693 - 23435745 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map