Assay Details
Target Gene Details
Entrez Gene ID: | 3030 |
Gene Name: | hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit |
Gene Aliases: |
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA |
Location: |
Chr.2:26190635-26244726 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HADHA | NM_000182.4 | NP_000173.2 | ||
| AK293398.1 | ||||
| AK299258.1 | ||||
| AK313027.1 | ||||
| BC009235.2 | AAH09235.1 | |||
| D16480.1 | BAA03941.1 | |||
| FJ626228.1 | ||||
| GU727642.1 | ||||
| U04627.1 | AAA56664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2648 | Chr.2:26177840 - 26222776 on Build GRCh38 | Deletion |
|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map