Assay Details
Target Gene Details
Entrez Gene ID: | 100129961 |
Gene Name: | CCNT2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:134866633-134918606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNT2-AS1 | NR_036549.1 | |||
| AK303295.1 |

Genomic Map