Assay Details
Target Gene Details
Entrez Gene ID: | 100379249 |
Gene Name: | BOK antisense RNA 1 |
Gene Aliases: |
BOK-AS, BOKAS, NAToB, NCRNA00151 |
Location: |
Chr.2:241544384-241559143 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BOK-AS1 | NR_033346.1 | |||
FJ185168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759120 | Chr.2:241480540 - 241562243 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv3584397 | Chr.2:241546796 - 241557859 on Build GRCh38 | Loss |
![]() |
nsv834588 | Chr.2:241498269 - 241613859 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
