Assay Details
Target Gene Details
Entrez Gene ID: | 55037 |
Gene Name: | pentatricopeptide repeat domain 3 |
Gene Aliases: |
MRP-S39 |
Location: |
Chr.2:86106182-86142157 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTCD3 | NM_017952.5 | NP_060422.4 | ||
| AK000765.1 | BAA91370.1 | |||
| AK293962.1 | ||||
| AK315676.1 | ||||
| AL832520.1 | CAH56345.1 | |||
| BC001758.1 | AAH01758.3 | |||
| BC011832.2 | AAH11832.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458563 | Chr.2:86065218 - 86280565 on Build GRCh38 | Gain |
|
| dgv3885n100 | Chr.2:86054844 - 86279009 on Build GRCh38 | Gain |
|
| dgv712e214 | Chr.2:86054752 - 86165643 on Build GRCh38 | Gain |
|
| esv2751903 | Chr.2:86026419 - 86278330 on Build GRCh38 | Gain |
|
| esv3591540 | Chr.2:86055590 - 86283809 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map