Assay Details
Target Gene Details
Entrez Gene ID: | 6233 |
Gene Name: | ribosomal protein S27a |
Gene Aliases: |
CEP80, HEL112, S27A, UBA80, UBC, UBCEP1, UBCEP80 |
Location: |
Chr.2:55231903-55235853 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS27A | NM_001135592.2 | NP_001129064.1 | ||
| NM_001177413.1 | NP_001170884.1 | |||
| NM_002954.5 | NP_002945.1 | |||
| AK093614.1 | 1 | 1395 | ||
| AK315695.1 | ||||
| BC042362.1 | AAH42362.1 | |||
| BF678026.1 | ||||
| BM907473.1 | ||||
| BQ276758.1 | ||||
| CK004647.1 | ||||
| CN344335.1 | ||||
| CR746519.1 | ||||
| DA527860.1 | ||||
| DB505702.1 | ||||
| EU794671.1 | ||||
| M10939.1 | AAA36788.1 | |||
| S79522.1 | AAB21188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2720078 | Chr.2:53962225 - 55281329 on Build GRCh38 | Deletion |
|
| nsv522016 | Chr.2:55177658 - 55333162 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map