Assay Details
Target Gene Details
Entrez Gene ID: | 100302650 |
Gene Name: | BRE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:27889456-27891114 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRE-AS1 | NR_028308.1 | 1 | 1391 | |
| AW080339.1 | 1 | 78 | ||
| DR978411.1 | ||||
| DR980668.1 |
Target Gene Details
Entrez Gene ID: | 64080 |
Gene Name: | ribokinase |
Gene Aliases: |
RBSK, RK |
Location: |
Chr.2:27781364-27890396 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBKS | NM_001287580.1 | NP_001274509.1 | ||
| NM_022128.2 | NP_071411.1 | |||
| AJ404857.1 | CAC12877.1 | |||
| AK300989.1 | ||||
| AY643715.1 | ||||
| BC017425.1 | AAH17425.1 | |||
| BX648909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759034 | Chr.2:27838469 - 28001763 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map