Assay Details
Target Gene Details
Entrez Gene ID: | 8837 |
Gene Name: | CASP8 and FADD like apoptosis regulator |
Gene Aliases: |
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS |
Location: |
Chr.2:201116015-201172688 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Gene Details
Entrez Gene ID: | 65072 |
Gene Name: | CFLAR antisense RNA 1 |
Gene Aliases: |
ALS2CR10 |
Location: |
Chr.2:201140289-201157792 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CFLAR-AS1 | NR_040030.1 | |||
AB053312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3593972 | Chr.2:201143394 - 201156199 on Build GRCh38 | Loss |
![]() ![]() |
nsv584160 | Chr.2:201039216 - 201298760 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
