Assay Details
Target Gene Details
Entrez Gene ID: | 57223 |
Gene Name: | protein phosphatase 4 regulatory subunit 3B |
Gene Aliases: |
FLFL2, PP4R3B, PSY2, SMEK2, smk1 |
Location: |
Chr.2:55547292-55617757 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575244 | Chr.2:55522427 - 56940881 on Build GRCh38 | Gain |
|
| esv2759050 | Chr.2:55520103 - 55753939 on Build GRCh38 | Gain |
|
| nsv1160801 | Chr.2:55524659 - 55588556 on Build GRCh38 | Duplication |
|
| nsv520581 | Chr.2:55395689 - 55675881 on Build GRCh38 | Loss |
|
| nsv525434 | Chr.2:55489514 - 55614504 on Build GRCh38 | Loss |
|
| nsv834080 | Chr.2:55513900 - 55700793 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72803546] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map