Assay Details
Target Gene Details
Entrez Gene ID: | 130106 |
Gene Name: | calcium and integrin binding family member 4 |
Gene Aliases: |
KIP4 |
Location: |
Chr.2:26581202-26641798 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIB4 | NM_001029881.2 | NP_001025052.1 | ||
| XM_011532514.2 | XP_011530816.1 | |||
| XM_011532517.2 | XP_011530819.1 | |||
| XM_017003329.1 | XP_016858818.1 | |||
| XM_017003331.1 | XP_016858820.1 | |||
| BC127684.1 | ||||
| BC127685.1 | ||||
| BC140923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517800 | Chr.2:26449146 - 26617624 on Build GRCh38 | Loss |
|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76585510,rs77806492] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map