Assay Details
Target Gene Details
Entrez Gene ID: | 344148 |
Gene Name: | NCK associated protein 5 |
Gene Aliases: |
ERIH1, ERIH2, NAP5 |
Location: |
Chr.2:132671788-133675181 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCKAP5 | NM_207363.2 | 20 | 7008 | NP_997246.2 |
| NM_207481.3 | 18 | 3051 | NP_997364.3 | |
| XM_005263659.3 | 18 | 7457 | XP_005263716.1 | |
| XM_005263660.3 | 18 | 6972 | XP_005263717.1 | |
| XM_011511097.2 | 20 | 7645 | XP_011509399.1 | |
| XM_011511098.2 | 20 | 7642 | XP_011509400.1 | |
| XM_011511099.2 | 19 | 7040 | XP_011509401.1 | |
| XM_011511100.2 | 19 | 7138 | XP_011509402.1 | |
| XM_011511101.1 | 17 | 6548 | XP_011509403.1 | |
| XM_011511102.2 | 18 | 10974 | XP_011509404.1 | |
| XM_011511103.2 | 16 | 6776 | XP_011509405.1 | |
| XM_011511104.1 | 16 | 6427 | XP_011509406.1 | |
| XM_011511105.2 | 16 | 7218 | XP_011509407.1 | |
| XM_017003974.1 | 19 | 6768 | XP_016859463.1 | |
| XM_017003975.1 | 18 | 7070 | XP_016859464.1 | |
| XM_017003976.1 | 20 | 7666 | XP_016859465.1 | |
| XM_017003977.1 | 17 | 7576 | XP_016859466.1 | |
| XM_017003978.1 | 13 | 6986 | XP_016859467.1 | |
| XM_017003979.1 | 17 | 3558 | XP_016859468.1 | |
| XM_017003980.1 | 15 | 2952 | XP_016859469.1 | |
| AK124659.1 | 7 | 1880 | BAC85920.1 | |
| CA438539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583158 | Chr.2:132558519 - 133104527 on Build GRCh38 | Gain |
|
| nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |
|
| nsv1011421 | Chr.2:132137668 - 132924612 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map