Assay Details
Target Gene Details
Entrez Gene ID: | 8440 |
Gene Name: | NCK adaptor protein 2 |
Gene Aliases: |
GRB4, NCKbeta |
Location: |
Chr.2:105744649-105894274 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCK2 | NM_001004722.3 | NP_001004722.1 | ||
| NM_003581.4 | NP_003572.2 | |||
| XM_006712797.3 | XP_006712860.1 | |||
| XM_011511991.2 | XP_011510293.1 | |||
| XM_017005103.1 | XP_016860592.1 | |||
| XM_017005105.1 | XP_016860594.1 | |||
| BG480643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834321 | Chr.2:105663236 - 105841053 on Build GRCh38 | Loss |
|
| dgv1159e212 | Chr.2:105767281 - 105773234 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map