Assay Details
Target Gene Details
Entrez Gene ID: | 102724691 |
Gene Name: | uncharacterized LOC102724691 |
Gene Aliases: |
- |
Location: |
Chr.2:104936241-105038496 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724691 | NR_110603.1 | |||
| BX109606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2862 | Chr.2:104945544 - 104980488 on Build GRCh38 | Insertion |
|

Genomic Map