Assay Details
Target Gene Details
Entrez Gene ID: | 23016 |
Gene Name: | exosome component 7 |
Gene Aliases: |
EAP1, RRP42, Rrp42p, hRrp42p, p8 |
Location: |
Chr.3:44975291-45012668 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOSC7 | NM_015004.3 | NP_055819.2 | ||
NR_023353.1 | ||||
XM_017005930.1 | XP_016861419.1 | |||
AK130053.1 | ||||
AK295779.1 | ||||
AK315741.1 | ||||
BC012831.2 | AAH12831.1 | |||
BU153856.1 | ||||
D29958.1 | BAA06226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834676 | Chr.3:44928707 - 45149356 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
