Assay Details
Target Gene Details
Entrez Gene ID: | 51517 |
Gene Name: | NCK interacting protein with SH3 domain |
Gene Aliases: |
AF3P21, DIP, DIP1, ORF1, SPIN90, VIP54, WASLBP, WISH |
Location: |
Chr.3:48663814-48685941 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCKIPSD | XM_017006594.1 | XP_016862083.1 | ||
| XM_017006595.1 | XP_016862084.1 | |||
| XM_017006596.1 | XP_016862085.1 | |||
| XM_017006597.1 | XP_016862086.1 | |||
| XM_017006598.1 | XP_016862087.1 | |||
| AJ242655.2 | CAB65089.2 | |||
| AY453794.1 | AAR83735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834685 | Chr.3:48527576 - 48708341 on Build GRCh38 | Loss |
|
| nsv523981 | Chr.3:48478880 - 48683607 on Build GRCh38 | Loss |
|
| nsv590247 | Chr.3:48624552 - 48764496 on Build GRCh38 | Loss |
|
| nsv818138 | Chr.3:48573492 - 48694054 on Build GRCh38 | Gain |
|
| nsv590246 | Chr.3:48590581 - 48683607 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78864109] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map